accessibility menu, dialog, popup

UserName

CAS RN 934987-26-5

IUPAC Name: 2-bromo-3,6-difluorobenzaldehyde
Synonyms: 2-bromo-3,6-difluorobenzaldehyde
Molecular Weight (g/mol): 221
Molecular Formula: C7H3BrF2O
InChi Key: PQXMBFZKKQNHBA-UHFFFAOYSA-N
SMILES: FC1=C(Br)C(C=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results