accessibility menu, dialog, popup

UserName

CAS RN 934570-45-3

CH3NNH2N

CAS RN 934570-45-3

IUPAC Name: 1-[3-(1-methyl-1H-pyrazol-5-yl)phenyl]methanamine
Synonyms: 1-[3-(2-methylpyrazol-3-yl)phenyl]methanamine
Molecular Weight (g/mol): 187.25
Molecular Formula: C11H13N3
InChi Key: UPQOTZRZQXMJNT-UHFFFAOYSA-N
SMILES: CN1N=CC=C1C1=CC(CN)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results