accessibility menu, dialog, popup

UserName

CAS RN 932-96-7

H3CNHCl

CAS RN 932-96-7

IUPAC Name: 4-chloro-N-methylaniline
Synonyms: 4-chlor-N-methylanilin
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: XCEYKKJMLOFDSS-UHFFFAOYSA-N
SMILES: CNC1=CC=C(Cl)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results