CAS RN 932-83-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitrosohexamethylenimine, TRC
CAS: 932-83-2 Molecular Formula: C6H12N2O Molecular Weight (g/mol): 128.17 Synonym: Hexahydro-1-nitroso-1H-azepine,N-Nitrosoazacycloheptane,N-Nitrosohexahydroazepine,Nitrosohexamethylenimine IUPAC Name: 1-nitrosoazepane SMILES: O=NN1CCCCCC1