accessibility menu, dialog, popup

UserName

CH3NHCH3N

CAS RN 930-62-1

IUPAC Name: 2,5-dimethyl-1H-imidazole
Synonyms: 2,4-dimethyl-3H-imidazole
Molecular Weight (g/mol): 96.13
Molecular Formula: C5H8N2
InChi Key: LLPKQRMDOFYSGZ-UHFFFAOYSA-N
SMILES: CC1=CN=C(C)N1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)