accessibility menu, dialog, popup

UserName

CAS RN 93-25-4

CH3OHOO

CAS RN 93-25-4

IUPAC Name: 2-(2-methoxyphenyl)acetic acid
Synonyms: benzeneacetic acid, 2-methoxy-
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: IVEWTCACRDEAOB-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1CC(O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)