accessibility menu, dialog, popup

UserName

HOBOHN

CAS RN 91983-26-5

IUPAC Name: [4-(cyanomethyl)phenyl]boronic acid
Synonyms: 4-(cyanomethyl)phenylboronic acid
Molecular Weight (g/mol): 160.97
Molecular Formula: C8H8BNO2
InChi Key: YKVMTTIYBSVPEQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(CC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results