accessibility menu, dialog, popup

UserName

ClHOFCl

CAS RN 916420-68-3

IUPAC Name: (3,6-dichloro-2-fluorophenyl)methanol
Synonyms: (3,6-dichloro-2-fluorophenyl)methanol
Molecular Weight (g/mol): 195.01
Molecular Formula: C7H5Cl2FO
InChi Key: VGTKYVUXLJVEPF-UHFFFAOYSA-N
SMILES: OCC1=C(Cl)C=CC(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results