accessibility menu, dialog, popup

UserName

CAS RN 916420-62-7

IUPAC Name: 3,6-dichloro-2-fluorobenzoic acid
Synonyms: 3,6-dichloro-2-fluorobenzoic acid
Molecular Weight (g/mol): 209
Molecular Formula: C7H3Cl2FO2
InChi Key: GPDYPRHGLQZRIP-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=CC(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results