accessibility menu, dialog, popup

UserName

CAS RN 916420-60-5

CH3OFFFNH2

CAS RN 916420-60-5

IUPAC Name: 1-[3-methyl-5-(trifluoromethoxy)phenyl]methanamine
Synonyms: 1-[3-methyl-5-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol): 205.18
Molecular Formula: C9H10F3NO
InChi Key: TXXSBJFLIMRWIO-UHFFFAOYSA-N
SMILES: CC1=CC(CN)=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results