accessibility menu, dialog, popup

UserName

BONHNOOHHO

CAS RN 913835-45-7

IUPAC Name: (4-{[2-(morpholin-4-yl)ethyl]carbamoyl}phenyl)boronic acid
Synonyms: 4-{[2-(morpholin-4-yl)ethyl]carbamoyl}phenylboronic acid
Molecular Weight (g/mol): 278.12
Molecular Formula: C13H19BN2O4
InChi Key: SFJAUBYTKBOMQG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C(=O)NCCN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results