CAS RN 91246-94-5
CAS RN 91246-94-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(1-hydroxy-1-phenylpropan-2-yl)-N-methylformamide, TRC
CAS: 91246-94-5 Molecular Formula: C11H15NO2 Molecular Weight (g/mol): 193.24 Synonym: N-(1-hydroxy-1-phenylpropan-2-yl)-N-methylformamide,rel-N-[(1R,2S)-2-Hydroxy-1-methyl-2-phenylethyl]-N-methylformamide IUPAC Name: N-(1-hydroxy-1-phenylpropan-2-yl)-N-methylformamide SMILES: CC(C(C1=CC=CC=C1)O)N(C)C=O