CAS RN 910297-51-7
CAS RN 910297-51-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Deshydroxyethyl Dasatinib, TRC
CAS: 910297-51-7 Molecular Formula: C20 H22 Cl N7 O S Molecular Weight (g/mol): 443.95 IUPAC Name: N-(2-chloro-6-methylphenyl)-2-[(2-methyl-6-piperazin-1-ylpyrimidin-4-yl)amino]-1,3-thiazole-5-carboxamide SMILES: Cc1cccc(Cl)c1NC(=O)c2cnc(Nc3cc(nc(C)n3)N4CCNCC4)s2