accessibility menu, dialog, popup

UserName

H3CONOO

CAS RN 91-23-6

IUPAC Name: 1-methoxy-2-nitrobenzene
Synonyms: 2-nitroanisole
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: CFBYEGUGFPZCNF-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)