accessibility menu, dialog, popup

UserName

CAS RN 906352-81-6

CH3HNNH3C

CAS RN 906352-81-6

IUPAC Name: methyl[(1-methyl-2,3-dihydro-1H-indol-5-yl)methyl]amine
Synonyms: methyl[(1-methyl-2,3-dihydroindol-5-yl)methyl]amine
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: QPUPWFNEDMKWGL-UHFFFAOYSA-N
SMILES: CNCC1=CC2=C(C=C1)N(C)CC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results