accessibility menu, dialog, popup

UserName

CAS RN 906352-73-6

NFFFNCO

CAS RN 906352-73-6

IUPAC Name: 2-(4-isocyanatophenyl)-5-(trifluoromethyl)pyridine
Synonyms: 2-(4-isocyanatophenyl)-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 264.21
Molecular Formula: C13H7F3N2O
InChi Key: GFEVPIXAPFROGH-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(C=C1)C1=CC=C(C=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results