accessibility menu, dialog, popup

UserName

CAS RN 906352-64-5

CH3HNNNN

CAS RN 906352-64-5

IUPAC Name: methyl({2-[(1H-1,2,4-triazol-1-yl)methyl]phenyl}methyl)amine
Synonyms: methyl({[2-(1,2,4-triazol-1-ylmethyl)phenyl]methyl})amine
Molecular Weight (g/mol): 202.26
Molecular Formula: C11H14N4
InChi Key: VHDVEBIJXKJALO-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1CN1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results