CAS RN 902454-26-6
IUPAC Name:
4-[(1-methylpiperidin-4-yl)methoxy]aniline
Synonyms:
4-[(1-methylpiperidin-4-yl)methoxy]aniline
Molecular Weight (g/mol):
220.32
Molecular Formula:
C13H20N2O
InChi Key:
WZRKMUXDLMLRJV-UHFFFAOYSA-N
SMILES:
CN1CCC(COC2=CC=C(N)C=C2)CC1
1
–
1
of
1
results
1
4-(1-Methyl-4-piperidinylmethoxy)aniline, 96%
CAS: 902454-26-6 Molecular Formula: C13H20N2O Molecular Weight (g/mol): 220.316 MDL Number: MFCD09909628 InChI Key: WZRKMUXDLMLRJV-UHFFFAOYSA-N Synonym: 4-1-methylpiperidin-4-yl methoxy aniline,4-1-methyl-4-piperidinylmethoxy aniline,4-1-methylpiperidin-4-yl methoxy phenylamine,4-1-methyl-piperidin-4-ylmethoxy-phenylamine,4-1-methyl-piperidin-4-ylmethoxy-phenyl amine,4-1-methyl-4-piperidinyl methoxy benzenamine PubChem CID: 11564798 IUPAC Name: 4-[(1-methylpiperidin-4-yl)methoxy]aniline SMILES: CN1CCC(CC1)COC2=CC=C(C=C2)N