CAS RN 9000-69-5
CAS RN 9000-69-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Pectin Citrus
CAS: 9000-69-5 MDL Number: MFCD00081838 Synonym: Poly-D-galacturonic acid methyl ester
2
Thermo Scientific Chemicals Pectin, pract.
CAS: 9000-69-5 MDL Number: MFCD00081838
3
Pectin, MP Biomedicals™
CAS: 9000-69-5 Molecular Formula: (C7H10O7)n Synonym: Poly-D-galacturonic acid methyl ester