accessibility menu, dialog, popup

UserName

CAS RN 90-14-2

IUPAC Name: 1-iodonaphthalene
Synonyms: 1-iodonaphthalene
Molecular Weight (g/mol): 254.07
Molecular Formula: C10H7I
InChi Key: NHPPIJMARIVBGU-UHFFFAOYSA-N
SMILES: IC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)