accessibility menu, dialog, popup

UserName

CAS RN 898544-48-4

IUPAC Name: potassium [2-(azidomethyl)phenyl]trifluoroboranuide
Synonyms: potassium [2-(azidomethyl)phenyl]trifluoroboranuide
Molecular Weight (g/mol): 239.05
Molecular Formula: C7H6BF3KN3
InChi Key: AKJNDNIFSIXTBR-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=CC=C1CN=[N+]=[N-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results