accessibility menu, dialog, popup

UserName

ONBrF

CAS RN 897016-96-5

IUPAC Name: 4-[(4-bromo-3-fluorophenyl)methyl]morpholine
Synonyms: 4-[(4-bromo-3-fluorophenyl)methyl]morpholine
Molecular Weight (g/mol): 274.13
Molecular Formula: C11H13BrFNO
InChi Key: AXLRZCPFGFYXIS-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC(CN2CCOCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results