accessibility menu, dialog, popup

UserName

CAS RN 892501-95-0

OOOH

CAS RN 892501-95-0

IUPAC Name: [4-(oxan-4-yloxy)phenyl]methanol
Synonyms: [4-(oxan-4-yloxy)phenyl]methanol
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: VFZJVDCLRTTYDL-UHFFFAOYSA-N
SMILES: OCC1=CC=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results