accessibility menu, dialog, popup

UserName

CAS RN 89205-07-2

IUPAC Name: 5-(4-methoxyphenyl)-1,3-oxazole-4-carboxylic acid
Synonyms: 5-(4-methoxyphenyl)-1,3-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 219.2
Molecular Formula: C11H9NO4
InChi Key: XVXCIDWYZMBWJU-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=C(N=CO1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results