accessibility menu, dialog, popup

UserName

CAS RN 89196-95-2

NNHOHCl

CAS RN 89196-95-2

IUPAC Name: 2-(1-benzofuran-2-yl)-4,5-dihydro-1H-imidazole hydrochloride
Synonyms: 2-(1-benzofuran-2-yl)-4,5-dihydro-1H-imidazole hydrochloride
Molecular Weight (g/mol): 222.67
Molecular Formula: C11H11ClN2O
InChi Key: RFNFFVDVGWOSNZ-UHFFFAOYSA-N
SMILES: Cl.C1CN=C(N1)C1=CC2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results