accessibility menu, dialog, popup

UserName

CH3HO

CAS RN 89-95-2

IUPAC Name: (2-methylphenyl)methanol
Synonyms: 2-methylbenzyl alcohol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: XPNGNIFUDRPBFJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results