accessibility menu, dialog, popup

UserName

NONO

CAS RN 88903-69-9

IUPAC Name: 9,30-dioxa-11,25-diazapentacyclo[20.6.2.2⁸,¹¹.0¹⁰,¹⁵.0²⁵,²⁹]dotriacontane
Synonyms: 9,30-dioxa-11,25-diazapentacyclo[20.6.2.2⁸,¹¹.0¹⁰,¹⁵.0²⁵,²⁹]dotriacontane
Molecular Weight (g/mol): 446.72
Molecular Formula: C28H50N2O2
InChi Key: PQYOPBRFUUEHRC-UHFFFAOYNA-N
SMILES: C1CC2CCCCCCC3CCN4CCCC(CCCCCCC5CCN(C1)C2O5)C4O3

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results