accessibility menu, dialog, popup

UserName

CAS RN 886851-30-5

CH3HNNNO

CAS RN 886851-30-5

IUPAC Name: methyl({[6-(morpholin-4-yl)pyridin-2-yl]methyl})amine
Synonyms: methyl({[6-(morpholin-4-yl)pyridin-2-yl]methyl})amine
Molecular Weight (g/mol): 207.28
Molecular Formula: C11H17N3O
InChi Key: CULRQHZBTPUFEB-UHFFFAOYSA-N
SMILES: CNCC1=NC(=CC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results