CAS RN 886766-60-5
IUPAC Name:
tert-butyl (2R)-2-phenylpiperazine-1-carboxylate
Synonyms:
tert-butyl (2R)-2-phenylpiperazine-1-carboxylate
Molecular Weight (g/mol):
262.35
Molecular Formula:
C15H22N2O2
InChi Key:
DVOURBIBCQYVCC-ZDUSSCGKSA-N
SMILES:
CC(C)(C)OC(=O)N1CCNC[C@H]1C1=CC=CC=C1
1
–
1
of
1
results
1
1-Boc-2-phenylpiperazine, 97%
CAS: 886766-60-5 Molecular Formula: C15H22N2O2 Molecular Weight (g/mol): 262.353 MDL Number: MFCD07367734 InChI Key: DVOURBIBCQYVCC-UHFFFAOYSA-N Synonym: 1-boc-2-phenylpiperazine,n-1-boc-2-phenyl piperizine,n-1-boc-2-phenylpiperazine,1-n-boc-2-phenylpiperazine,n-boc-2-bhenyl piperazine,2-phenylpiperazine, n1-boc protected,2-phenyl-piperazine-1-carboxylic acid tert-butyl ester,n1-boc-2-phenylpiperazine,n-1-boc-2-phenyl-piperazine,tert-butyl 2-phenylpiperazinecarboxylate PubChem CID: 16740572 IUPAC Name: tert-butyl 2-phenylpiperazine-1-carboxylate SMILES: CC(C)(C)OC(=O)N1CCNCC1C2=CC=CC=C2