accessibility menu, dialog, popup

UserName

CAS RN 886761-75-7

H3COClONH2F

CAS RN 886761-75-7

IUPAC Name: 6-chloro-2-fluoro-3-methoxybenzamide
Synonyms: 6-chloro-2-fluoro-3-methoxybenzamide
Molecular Weight (g/mol): 203.6
Molecular Formula: C8H7ClFNO2
InChi Key: AZUOJQIOSGRMMS-UHFFFAOYSA-N
SMILES: COC1=C(F)C(C(N)=O)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results