CAS RN 886761-66-6
IUPAC Name:
2-(3-chloro-2,4-difluorophenyl)acetic acid
Synonyms:
(3-chloro-2,4-difluorophenyl)acetic acid
Molecular Weight (g/mol):
206.57
Molecular Formula:
C8H5ClF2O2
InChi Key:
HENNVWWQJGUCAH-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=C(F)C(Cl)=C(F)C=C1
1
–
1
of
1
results
1
3-Chloro-2,4-difluorophenylacetic acid, 97%, Thermo Scientific™
CAS: 886761-66-6 Molecular Formula: C8H5ClF2O2 Molecular Weight (g/mol): 206.573 MDL Number: MFCD06660337 InChI Key: HENNVWWQJGUCAH-UHFFFAOYSA-N Synonym: 3-chloro-2,4-difluorophenylacetic acid,3-chloro-2,4-difluorophenyl acetic acid,2-3-chloro-2,4-difluorophenyl acetic acid,benzeneacetic acid, 3-chloro-2,4-difluoro PubChem CID: 17750824 IUPAC Name: 2-(3-chloro-2,4-difluorophenyl)acetic acid SMILES: C1=CC(=C(C(=C1CC(=O)O)F)Cl)F