accessibility menu, dialog, popup

UserName

CAS RN 886499-13-4

IUPAC Name: 1-[3-fluoro-4-(trifluoromethoxy)phenyl]methanamine
Synonyms: 1-[3-fluoro-4-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol): 209.14
Molecular Formula: C8H7F4NO
InChi Key: LKKOOHKPOMOOAI-UHFFFAOYSA-N
SMILES: NCC1=CC(F)=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results