CAS RN 886498-75-5
CAS RN 886498-75-5
IUPAC Name:
3,5-difluoro-2-methoxybenzoic acid
Synonyms:
3,5-difluoro-2-methoxybenzoic acid
Molecular Weight (g/mol):
188.13
Molecular Formula:
C8H6F2O3
InChi Key:
MEVHLVDNBMQMNH-UHFFFAOYSA-N
SMILES:
COC1=C(F)C=C(F)C=C1C(O)=O
1
–
1
of
1
results
1
3,5-Difluoro-2-methoxybenzoic acid, 97%, Thermo Scientific™
CAS: 886498-75-5 Molecular Formula: C8H6F2O3 Molecular Weight (g/mol): 188.13 MDL Number: MFCD06660198 InChI Key: MEVHLVDNBMQMNH-UHFFFAOYSA-N Synonym: pubchem4942,3,5-difluoro-2-methoxybenzoicacid,3,5-difluoro-2-methoxy-benzoic acid PubChem CID: 17750763 IUPAC Name: 3,5-difluoro-2-methoxybenzoic acid SMILES: COC1=C(C=C(C=C1C(=O)O)F)F