CAS RN 886497-98-9
CAS RN 886497-98-9
IUPAC Name:
2-[2-fluoro-5-(trifluoromethoxy)phenyl]acetic acid
Synonyms:
[2-fluoro-5-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol):
238.14
Molecular Formula:
C9H6F4O3
InChi Key:
PYCWSPHSXHTRQT-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=C(F)C=CC(OC(F)(F)F)=C1
1
–
1
of
1
results
1
2-Fluoro-5-(trifluoromethoxy)phenylacetic acid, 97%, Thermo Scientific™
CAS: 886497-98-9 Molecular Formula: C9H6F4O3 Molecular Weight (g/mol): 238.14 MDL Number: MFCD04115892 InChI Key: PYCWSPHSXHTRQT-UHFFFAOYSA-N Synonym: 2-fluoro-5-trifluoromethoxy phenylacetic acid,2-2-fluoro-5-trifluoromethoxy phenyl acetic acid,2-fluoro-5-trifluoromethoxy phenyl acetic acid PubChem CID: 2783328 IUPAC Name: 2-[2-fluoro-5-(trifluoromethoxy)phenyl]acetic acid SMILES: OC(=O)CC1=C(F)C=CC(OC(F)(F)F)=C1