accessibility menu, dialog, popup

UserName

BrSH

CAS RN 886497-84-3

IUPAC Name: (3-bromophenyl)methanethiol
Synonyms: (3-bromophenyl)methanethiol
Molecular Weight (g/mol): 203.1
Molecular Formula: C7H7BrS
InChi Key: LXEHTZZVGCUFCS-UHFFFAOYSA-N
SMILES: SCC1=CC(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results