accessibility menu, dialog, popup

UserName

CAS RN 884494-44-4

NIHOCl

CAS RN 884494-44-4

IUPAC Name: (2-chloro-4-iodopyridin-3-yl)methanol
Synonyms: (2-chloro-4-iodopyridin-3-yl)methanol
Molecular Weight (g/mol): 269.47
Molecular Formula: C6H5ClINO
InChi Key: BZQRQPAKTCMYIA-UHFFFAOYSA-N
SMILES: OCC1=C(I)C=CN=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results