CAS RN 88378-62-5
CAS RN 88378-62-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(17Beta)-Estra-1,3,5(10),6-tetraene-3,17Beta-diol 17-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(17Beta)-Estra-1,3,5(10),6-tetraene-3,17Beta-diol 17-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(17b)-Estra-1,3,5(10),6-tetraene-3,17b-diol 17-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.