CAS RN 881-07-2
CAS RN 881-07-2
IUPAC Name:
2-methyl-8-nitroquinoline
Synonyms:
quinoline, 2-methyl-8-nitro-
Molecular Weight (g/mol):
188.19
Molecular Formula:
C10H8N2O2
InChi Key:
UHPGVDHXHDPYQP-UHFFFAOYSA-N
SMILES:
CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O
1
–
1
of
1
results
1
2-Methyl-8-nitroquinoline, 98%
CAS: 881-07-2 Molecular Formula: C10H8N2O2 Molecular Weight (g/mol): 188.19 MDL Number: MFCD00006764 InChI Key: UHPGVDHXHDPYQP-UHFFFAOYSA-N PubChem CID: 13433 IUPAC Name: 2-methyl-8-nitroquinoline SMILES: CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O