accessibility menu, dialog, popup

UserName

CAS RN 88-44-8

IUPAC Name: 2-amino-5-methylbenzene-1-sulfonic acid
Synonyms: 2-amino-5-methylbenzenesulfonic acid
Molecular Weight (g/mol): 187.21
Molecular Formula: C7H9NO3S
InChi Key: LTPSRQRIPCVMKQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(=C1)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results