accessibility menu, dialog, popup

UserName

CAS RN 88-21-1

NH2SHOOO

CAS RN 88-21-1

IUPAC Name: 2-aminobenzene-1-sulfonic acid
Synonyms: orthanilic acid o-sulfanilic acid O-anilinesulfonic acid 2-sulfanilic acid
Molecular Weight (g/mol): 173.19
Molecular Formula: C6H7NO3S
InChi Key: ZMCHBSMFKQYNKA-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1S(O)(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)