accessibility menu, dialog, popup

UserName

CAS RN 877-22-5

CH3OOHOHO

CAS RN 877-22-5

IUPAC Name: 2-hydroxy-3-methoxybenzoic acid
Synonyms: 3-methoxysalicyclic acid
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: AUZQQIPZESHNMG-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C(O)=O)=C1O
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results