accessibility menu, dialog, popup

UserName

H3CONCH3OCH3NHOOCH3CH3H3C

CAS RN 87694-49-3

IUPAC Name: tert-butyl N-{1-[methoxy(methyl)carbamoyl]ethyl}carbamate
Synonyms: tert-butyl N-{1-[methoxy(methyl)carbamoyl]ethyl}carbamate
Molecular Weight (g/mol): 232.28
Molecular Formula: C10H20N2O4
InChi Key: PWQIGBOSLQHOBT-UHFFFAOYNA-N
SMILES: CON(C)C(=O)C(C)NC(=O)OC(C)(C)C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)