accessibility menu, dialog, popup

UserName

H3COSOONH2F

CAS RN 874767-60-9

IUPAC Name: 3-fluoro-4-methoxybenzene-1-sulfonamide
Synonyms: 3-fluoro-4-methoxybenzenesulfonamide
Molecular Weight (g/mol): 205.2
Molecular Formula: C7H8FNO3S
InChi Key: JCPWNWBBZNNHDW-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(C=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results