accessibility menu, dialog, popup

UserName

HOBOHFHOO

CAS RN 874219-35-9

IUPAC Name: 5-(dihydroxyboranyl)-2-fluorobenzoic acid
Synonyms: 5-(dihydroxyboranyl)-2-fluorobenzoic acid
Molecular Weight (g/mol): 183.93
Molecular Formula: C7H6BFO4
InChi Key: DGORTXQWDDXSIQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(F)C(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results