accessibility menu, dialog, popup

UserName

H3COCH3H3C

CAS RN 874-63-5

IUPAC Name: 1-methoxy-3,5-dimethylbenzene
Synonyms: 3,5-dimethylanisole
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: JCHJBEZBHANKGA-UHFFFAOYSA-N
SMILES: COC1=CC(C)=CC(C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)