accessibility menu, dialog, popup

UserName

CAS RN 872-55-9

H3CS

CAS RN 872-55-9

IUPAC Name: 2-ethylthiophene
Synonyms: 2-ethylthiophene
Molecular Weight (g/mol): 112.19
Molecular Formula: C6H8S
InChi Key: JCCCMAAJYSNBPR-UHFFFAOYSA-N
SMILES: CCC1=CC=CS1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)