accessibility menu, dialog, popup

UserName

BClOHNCH3HOOH

CAS RN 871332-65-9

IUPAC Name: [4-chloro-3-(methylcarbamoyl)phenyl]boronic acid
Synonyms: 4-chloro-3-(methylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 213.42
Molecular Formula: C8H9BClNO3
InChi Key: SYZZWFMATUWETK-UHFFFAOYSA-N
SMILES: CNC(=O)C1=C(Cl)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results