accessibility menu, dialog, popup

UserName

CAS RN 87031-30-9

CH3H3CCH3NNOCH3

CAS RN 87031-30-9

IUPAC Name: 3-tert-butyl-1-methyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 5-tert-butyl-2-methyl-4H-pyrazol-3-one
Molecular Weight (g/mol): 154.21
Molecular Formula: C8H14N2O
InChi Key: QTLHXZDNJCSMMQ-UHFFFAOYSA-N
SMILES: CN1N=C(CC1=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results