accessibility menu, dialog, popup

UserName

CAS RN 86873-60-1

NClOHO

CAS RN 86873-60-1

IUPAC Name: 5-chloropyridine-2-carboxylic acid
Synonyms: 5-chloropyridine-2-carboxylic acid
Molecular Weight (g/mol): 157.55
Molecular Formula: C6H4ClNO2
InChi Key: GJLOKYIYZIOIPN-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results