accessibility menu, dialog, popup

UserName

H3COO(S)OHCl

CAS RN 86728-85-0

IUPAC Name: ethyl (3S)-4-chloro-3-hydroxybutanoate
Synonyms: ethyl (3S)-4-chloro-3-hydroxybutanoate
Molecular Weight (g/mol): 166.6
Molecular Formula: C6H11ClO3
InChi Key: ZAJNMXDBJKCCAT-YFKPBYRVSA-N
SMILES: CCOC(=O)C[C@H](O)CCl
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)